ContactUsLinkComponent
Liquid electrolyte comb. electrode with Pt 1000 and integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum ju nction, electrolyte L 200, sphere membrane, A-glass, VP screw plug head, length 225 mm, 12 mm Ø, -30...+100°C, 0...14 1 * 1 items
Catalog # SCOI285110310
Supplier:  SI Analytics
Liquid electrolyte comb. electrode with Pt 1000 and integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum ju nction, electrolyte L 200, sphere membrane, A-glass, VP screw plug head, length 225 mm, 12 mm Ø, -30...+100°C, 0...14 1 * 1 items
Catalog # SCOI285110310
Supplier:  SI Analytics