ContactUsLinkComponent
One hose coupling for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123240
Supplier:  SI Analytics
One hose coupling for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123240
Supplier:  SI Analytics