CYCLOHEXENE, INHIBITOR-FREE, REAGENTPLU 1 * 100 mL
Catalog # SIAL125431-100ML
Supplier: Merck
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzylacetát for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitril 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
4-Methylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
1-Butanol NF
Supplier: Spectrum Chemical
Isooktán 99.5% p.a.
Supplier: Thermo Fisher Scientific
[EN]TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
izo-Amylalkohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Supplier: PanReac AppliChem