ContactUsLinkComponent
FITC-CM-Dextran, average mol wt 40,000 1 * 1 g
Catalog # SIAL53379-1G
Supplier:  Merck
FITC-CM-Dextran, average mol wt 40,000 1 * 1 g
Catalog # SIAL53379-1G
Supplier:  Merck
Frequently Bought Together