cis-11-Methyl-2-dodecenoic acid 1 * 10 mg
SIAL42052-10MG
: Merck
Frequently Bought Together
E.Z.N.A.® Plasmid Mini kit I
: OMEGA BIO-TEK
Phenetole ≥98%
: Thermo Fisher Scientific
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
: Thermo Fisher Scientific
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade
: Honeywell Chemicals
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
: MACRON AVANTOR BRAND
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
: Thermo Fisher Scientific
Methanol 99.9% for analysis
: Thermo Fisher Scientific