SODIUM BROMIDE, ANHYDROUS, FREE-FLOWING 1 * 500 g
Catalog # SIAL793574-500G
Supplier: Merck
Frequently Bought Together
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Silver nitrate 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Magnesium chloride hexahydrate 98.0-101.0% USP, Ph. Eur., BP
Supplier: VWR Chemicals
Potassium fluoride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals