ContactUsLinkComponent
Carrier bags are suitable for convenient transport of one or several vacuette® transport boxes or containers. the transport bags con sist of isothermal material and are thus equally suitable for keeping samples cool. the thermoscan temperature monito 1 * 1 items
Catalog # GREI472022_1
Supplier:  Greiner Bio-One
Carrier bags are suitable for convenient transport of one or several vacuette® transport boxes or containers. the transport bags con sist of isothermal material and are thus equally suitable for keeping samples cool. the thermoscan temperature monito 1 * 1 items
Catalog # GREI472022_1
Supplier:  Greiner Bio-One
Frequently Bought Together