ContactUsLinkComponent
Coagulation and ctad tubes are used for examination of coagulation parameters. coagulation tubes contain a buffered sodium citrate s olution. it is available with a citrate concentration of 0.109 mol/l (3.2%) or 0.129 mol/l (3.8%). the mixing ratio i 1 * 1.200 items
Catalog # GREI454064_1200
Supplier:  Greiner Bio-One
Coagulation and ctad tubes are used for examination of coagulation parameters. coagulation tubes contain a buffered sodium citrate s olution. it is available with a citrate concentration of 0.109 mol/l (3.2%) or 0.129 mol/l (3.8%). the mixing ratio i 1 * 1.200 items
Catalog # GREI454064_1200
Supplier:  Greiner Bio-One
Frequently Bought Together