ContactUsLinkComponent
Flask, round bottom, 500 ml, two side necks at an angle of 20, center neck ST 29/32, side neck ST 14/23, acc. to DIN 12394 1 * 1 items
Catalog # WITG0.736.500
Supplier:  witeg Labortechnik
Flask, round bottom, 500 ml, two side necks at an angle of 20, center neck ST 29/32, side neck ST 14/23, acc. to DIN 12394 1 * 1 items
Catalog # WITG0.736.500
Supplier:  witeg Labortechnik
Frequently Bought Together