ContactUsLinkComponent
Heating module accessory for MicroSolvCE Window Maker. Makes approximately a 1mm burn area. Use in Capillary Electrophoresis. 1 * 1 items
Catalog # MICS07301-S
Supplier:  MICROSOLV
undefined
Heating module accessory for MicroSolvCE Window Maker. Makes approximately a 1mm burn area. Use in Capillary Electrophoresis. 1 * 1 items
Catalog # MICS07301-S
Supplier:  MICROSOLV
Frequently Bought Together