ContactUsLinkComponent
7 UV Calibration Test Replacement Absorbance Solutions. 1 Wavelength Solution, 1 Stray Light Solution, 1 Reference Solution. Certifi cate of Analysis. 1 * 7 Bottle
Catalog # MICS49115-25
Supplier:  MICROSOLV
undefined
7 UV Calibration Test Replacement Absorbance Solutions. 1 Wavelength Solution, 1 Stray Light Solution, 1 Reference Solution. Certifi cate of Analysis. 1 * 7 Bottle
Catalog # MICS49115-25
Supplier:  MICROSOLV
Frequently Bought Together