ContactUsLinkComponent
NIST Traceable test solutions kit for use with MicroSolv HSQ HPLC PQ performance qualification test kit with fluorescence detectors. 6x30ml salicylic acid and 1x30ml blank diliuent soltuions w/certificate of analysis. Supplied with 1 Excel template s 1 * 30 mL
Catalog # MICS49103-FL
Supplier:  MICROSOLV
undefined
NIST Traceable test solutions kit for use with MicroSolv HSQ HPLC PQ performance qualification test kit with fluorescence detectors. 6x30ml salicylic acid and 1x30ml blank diliuent soltuions w/certificate of analysis. Supplied with 1 Excel template s 1 * 30 mL
Catalog # MICS49103-FL
Supplier:  MICROSOLV
Frequently Bought Together