ContactUsLinkComponent
Easy Purchase Pack. MRQ Maximum Recovery screw top autosampler vials made with RSA Glass, clear 1.2ml fill volume, 12x32mm and poly ethylene, single injection caps. Clear tray pack. 100/pack. 1 * 100 items
Catalog # MICS9512S-9CP-T-RS
Supplier:  MICROSOLV
undefined
Easy Purchase Pack. MRQ Maximum Recovery screw top autosampler vials made with RSA Glass, clear 1.2ml fill volume, 12x32mm and poly ethylene, single injection caps. Clear tray pack. 100/pack. 1 * 100 items
Catalog # MICS9512S-9CP-T-RS
Supplier:  MICROSOLV
Frequently Bought Together