ContactUsLinkComponent
Easy Purchase Pack, 2ml amber vials with RSA-Pro glass treatment (12x32mm) with write on patch and light blue AQR screw caps with pr e-slit fitted Silicone/PTFE septa. RSA-Pro Brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-3WDAP-MR
Supplier:  MICROSOLV
undefined
Easy Purchase Pack, 2ml amber vials with RSA-Pro glass treatment (12x32mm) with write on patch and light blue AQR screw caps with pr e-slit fitted Silicone/PTFE septa. RSA-Pro Brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-3WDAP-MR
Supplier:  MICROSOLV
Frequently Bought Together