ContactUsLinkComponent
Easy Purchase Pack, 2ml clear vials with RSA-Pro glass treatment (12x32mm) w/ write on patch and light blue AQR screw caps with fitt ed silicone/PTFE septa. AQ Brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-1WDCP-MR
Supplier:  MICROSOLV
undefined
Easy Purchase Pack, 2ml clear vials with RSA-Pro glass treatment (12x32mm) w/ write on patch and light blue AQR screw caps with fitt ed silicone/PTFE septa. AQ Brand. 100/pack. 1 * 100 items
Catalog # MICS9509S-1WDCP-MR
Supplier:  MICROSOLV
Frequently Bought Together