ContactUsLinkComponent
Caps, screw top with single slit, Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Red, for use with 9-425mm, 12x32m m autosampler vials. BASIK brand. 1,000/case 1 * 1.000 items
Catalog # MICS9502S-E-30C-RM
Supplier:  MICROSOLV
undefined
Caps, screw top with single slit, Soft-Guard white silicone/ red PTFE septa, bonded in ribbed cap. Red, for use with 9-425mm, 12x32m m autosampler vials. BASIK brand. 1,000/case 1 * 1.000 items
Catalog # MICS9502S-E-30C-RM
Supplier:  MICROSOLV
Frequently Bought Together