ContactUsLinkComponent
Test sieve stainless steel with wire gauze 450 mm Ø x 100 mm height ASTM E 11 1.00 mm 18 Mesh 1 * 1 items
Catalog # RETS60.169.001000
Supplier:  Retsch GmbH
Test sieve stainless steel with wire gauze 450 mm Ø x 100 mm height ASTM E 11 1.00 mm 18 Mesh 1 * 1 items
Catalog # RETS60.169.001000
Supplier:  Retsch GmbH
Frequently Bought Together