ContactUsLinkComponent
Probe 1 (25,4 mm) for more difficult applications and larger volumes (2 - 20 L) 1 * 1 items
Catalog # QSON4617
Supplier:  QSONICA LLC
Probe 1 (25,4 mm) for more difficult applications and larger volumes (2 - 20 L) 1 * 1 items
Catalog # QSON4617
Supplier:  QSONICA LLC
Frequently Bought Together