CLEANING CONCENTRATE ELMA LAB CLEAN S15 1 * 25 L
Catalog # ELMA8000432
Supplier: ELMA SCHMIDBAUER
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Membrane filters with grid
Supplier: Sartorius
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dip slides for surfaces and in liquids, Envirocheck®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
LAL chromogenic endotoxin assay kit, ToxinSensor™
Supplier: Genscript
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals