ContactUsLinkComponent
VAC MODEL CR-1000(CV) 230V w/EURO PLUG w/SAFE CONTAINMENT - TOOL KIT SOLD SEPARATELY Technical features: Volt: 240V, Watt:1000 W Hertz: 50 Hz Ampere: 4.3 A. Dry recovery. Air Flow: 163 m /h Vacuum pressure: 2515 mmH20 Containment Recovery: 7.6L D 1 * 1 items
Catalog # TIGV111993B
Supplier:  TIGER-VAC
VAC MODEL CR-1000(CV) 230V w/EURO PLUG w/SAFE CONTAINMENT - TOOL KIT SOLD SEPARATELY Technical features: Volt: 240V, Watt:1000 W Hertz: 50 Hz Ampere: 4.3 A. Dry recovery. Air Flow: 163 m /h Vacuum pressure: 2515 mmH20 Containment Recovery: 7.6L D 1 * 1 items
Catalog # TIGV111993B
Supplier:  TIGER-VAC
Frequently Bought Together