ContactUsLinkComponent
VACUUM MODEL SS-15(TC)TE w/TANGENTIAL EXHAUST FOR FUEL, SOVLENT AND OIL RECOVERY Wet Recovery: 47L Vacuum Pressure: 5080 mmH20 Air Flow: 187m /h Dimensions: H97 cm x W69 cm x L89 cm Weight: 38 Kg Noise level: 70.00 dB(A) 1 * 1 items
Catalog # TIGV110270
Supplier:  TIGER-VAC
VACUUM MODEL SS-15(TC)TE w/TANGENTIAL EXHAUST FOR FUEL, SOVLENT AND OIL RECOVERY Wet Recovery: 47L Vacuum Pressure: 5080 mmH20 Air Flow: 187m /h Dimensions: H97 cm x W69 cm x L89 cm Weight: 38 Kg Noise level: 70.00 dB(A) 1 * 1 items
Catalog # TIGV110270
Supplier:  TIGER-VAC
Frequently Bought Together