ContactUsLinkComponent
Agilent J and W GC packed column in UltiMetal tubing, length 4.92 ft (1.5 m), 1/4 inch od, 4 mm id, Hayesep R packing, mesh size 60/ 80, preconditioned 1 * 1 items
  HEWLG3591-81185
 :  Agilent
Agilent J and W GC packed column in UltiMetal tubing, length 4.92 ft (1.5 m), 1/4 inch od, 4 mm id, Hayesep R packing, mesh size 60/ 80, preconditioned 1 * 1 items
  HEWLG3591-81185
 :  Agilent
Frequently Bought Together