ContactUsLinkComponent
ICH/USP oral target elements standard D: Cr 11,000 µg/mL, Sn 6000 µg/mL, Cu and Mo 3000 µg/mL, Ba 1400 µg/mL, Sb 1200 µg/mL, Li 550 µg/mL in 5% HNO3/tr. HF 1 * 1 items
Catalog # HEWL5190-9769
Supplier:  Agilent
undefined
ICH/USP oral target elements standard D: Cr 11,000 µg/mL, Sn 6000 µg/mL, Cu and Mo 3000 µg/mL, Ba 1400 µg/mL, Sb 1200 µg/mL, Li 550 µg/mL in 5% HNO3/tr. HF 1 * 1 items
Catalog # HEWL5190-9769
Supplier:  Agilent
Frequently Bought Together