ContactUsLinkComponent
Method development capilary kit, includes capillaries and Quick Connect heat exchangers to support the installation of up to 8 long columns distributed over 2 multicolumn thermostats with one 8 column selector valve 1 * 1 items
Catalog # HEWL5067-6697
Supplier:  Agilent
Method development capilary kit, includes capillaries and Quick Connect heat exchangers to support the installation of up to 8 long columns distributed over 2 multicolumn thermostats with one 8 column selector valve 1 * 1 items
Catalog # HEWL5067-6697
Supplier:  Agilent
Frequently Bought Together