ContactUsLinkComponent
BrightMax™ sealing films are 200 µm thick white vinyl adhesive sealing films for use with white microplates. The sealing films may be applied to either the top or bottom of the plate. A split backing aids in applying the sealing film properly. The li 1 * 50 items
Catalog # SIALZ732117-50EA
Supplier:  Merck
BrightMax™ sealing films are 200 µm thick white vinyl adhesive sealing films for use with white microplates. The sealing films may be applied to either the top or bottom of the plate. A split backing aids in applying the sealing film properly. The li 1 * 50 items
Catalog # SIALZ732117-50EA
Supplier:  Merck
Frequently Bought Together