ContactUsLinkComponent
AbsorbMax films (76,2 mm × 114,3 mm) are 200 µm thick black vinyl non-sterile adhesive films for use with 96 well, 384 well, and 1536 well black microplates. The films may be applied to either the top or bottom of the plate. A split backing aids in a 1 * 50 items
Catalog # SIALZ722537-50EA
Supplier:  Merck
AbsorbMax films (76,2 mm × 114,3 mm) are 200 µm thick black vinyl non-sterile adhesive films for use with 96 well, 384 well, and 1536 well black microplates. The films may be applied to either the top or bottom of the plate. A split backing aids in a 1 * 50 items
Catalog # SIALZ722537-50EA
Supplier:  Merck
Frequently Bought Together