Eppendorf® Pipette Holder System
Product Details & Documents
Save valuable bench space with the Eppendorf Pipette Holder System! With space for up to six single- or multi-channel pipettes, you can easily store all of your Eppendorf pipettes and multi-dispensers within easy reach. Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors.
- The Eppendorf pipette holder system stores all current mechanical and electronic Eppendorf pipettes (including the Repeater®) and most of their predecessors
- Pipette Carousel 2: Rotatable holder with completely new design for all mechanical pipettes including Eppendorf Research®, Research plus, Reference, Reference 2 and Biomaster®
- Charer Carousel 2: Carousel with intelligent charging electronics and increased capacity for up to six instruments (charges four instruments at a time and all six in turn)
- Carousels and stands with interchangeable pipette holders and charger shells, no tools required
- Large rubber feet protect carousels and stands from liquids spilled on bench top
- Pictograms on all charger shells and pipette holders for clear assignment in the lab
Well within reach: The Eppendorf pipette holder system helps you to organise your lab.
The robust Eppendorf pipette holder system allows you to store up to six of your Eppendorf pipettes or hand dispensers safely and securely in a small footprint, freeing up valuable bench space. Pipettes are positioned upright to prevent their nose cones from coming into contact with work surfaces and prevent damage and risk of cross-contamination. Pipette carousels and stands give you easy access to your instrument of choice, and are available with or without a charging station. Pipette stands carry one electronic pipette or Repeater®, and wall mount devices store one mechanical or electronic pipette, or Repeater.
Ordering information: Includes charger shells for six Eppendorf Xplorer®/Xplorer® plus and holders for 6 Eppendorf Research®, Eppendorf Research® plus, Eppendorf Reference® 2.
Documents
Eppendorf Pipette Holder System
Product Literature
Specifications
- Description:Charger stand 2, for 1 Eppendorf Xplorer® or Xplorer® plus
- Seq:02
- Cat. No.:EPPE3116000.031
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact