ContactUsLinkComponent
DMEM:F-12 for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS). 1 * 500 mL
Catalog # PIER88370
DMEM:F-12 for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS). 1 * 500 mL
Catalog # PIER88370
Frequently Bought Together