MEMOSENS PH SENSOR SE515 L 120MM 1 * 1 KS
KNICSE515/1-MS
: KNICK
Frequently Bought Together
VWR®, Sérologická pipeta, standardní
: VWR Collection
Transfer pipettes
: RATIOLAB
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
pH buffer solutions, NIST/DIN
: Mettler - Toledo
Ethanol 96% vol Ph. Eur.
: VWR Chemicals