ContactUsLinkComponent
IL2RA (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of IL2RA expression in cul tured cells. 1 * 1 KIT
Catalog # ABNOKA2789
Supplier:  Abnova
IL2RA (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of IL2RA expression in cul tured cells. 1 * 1 KIT
Catalog # ABNOKA2789
Supplier:  Abnova
Frequently Bought Together