ContactUsLinkComponent
TFRC (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of TFRC expression in cultu red cells. 1 * 1 KIT
Catalog # ABNOKA2658
Supplier:  Abnova
TFRC (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of TFRC expression in cultu red cells. 1 * 1 KIT
Catalog # ABNOKA2658
Supplier:  Abnova
Frequently Bought Together