FEXOFENADINE HCL 1 * 1 items
Catalog # PROOMM0507.00
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Ethanolamine, Supelco®
Supplier: Merck
GYPENOSIDE XVII, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
SILDENAFIL 1 * 10 mg
Supplier: LGC Standards PROMOCHEM
VWR®, Sulphur, Certified Reference Materials, in light mineral oil
Supplier: VWR Chemicals
Sodium standard solution, 1000 mg/L Na in 5% nitric acid (from Na₂CO₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
HEXACHLOROBENZENE-13C6 100G/ML IN ACET 1 * 1 mL
Supplier: RIVOLAB
ISE standards
Supplier: Hach
Trifluralin D14 (di-n-propyl D14) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
Maduramicin ammonium salt certified 1 * 0,1 g
Supplier: EHRENSTORFER
VWR®, Succinate 1000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, Octanal
Supplier: Merck
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
Reagents for chlorine determination using Lovibond® MD photometers
Supplier: Lovibond