ContactUsLinkComponent
DESMOGLEIN 4 POLYCLONAL ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
  BOSSBS-13744R-A750
 :  Bioss
DESMOGLEIN 4 POLYCLONAL ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
  BOSSBS-13744R-A750
 :  Bioss
Frequently Bought Together