Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Wire baskets, rectangular, with lid

80×40×30 mm
  HUPF7501282
Wire baskets, rectangular, with lid
  HUPF7501282
Wire baskets, rectangular, with lid
Wire baskets, rectangular, with lid

80×40×30 mm

 
5 770,00 Kč
   
1 * 1 KS
 

These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.

  • With lid
  • With base frame
  • Mesh size: 1,6×1,6 mm

Colour: Silver.

 
  • Seq:
    01
  • W×D×H:
    80×40×30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.