0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
H1N1 HEMAGGLUTININ 1 POLYCLONAL ANTIBODY, ALEXA FLUOR 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: VIRUS, APPLICATION: IF( IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 l
BOSSBS-2001R-A750
:
Bioss
H1N1 HEMAGGLUTININ 1 POLYCLONAL ANTIBODY, ALEXA FLUOR 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: VIRUS, APPLICATION: IF( IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 l
BOSSBS-2001R-A750
:
Bioss
About BIOSS
bs-2001R-A750
Frequently Bought Together
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Reag. USP, ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection