ContactUsLinkComponent
DYNORPHIN A/PROENKEPHALIN B ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICAT ION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-13041R-A750
Supplier:  Bioss
DYNORPHIN A/PROENKEPHALIN B ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICAT ION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-13041R-A750
Supplier:  Bioss