0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
PDMS, viscoelastic material with certified cross-over (20 C) 1 * 50 mL
HAAK082-5491
:
HAAKE
PDMS, viscoelastic material with certified cross-over (20 C) 1 * 50 mL
HAAK082-5491
:
HAAKE
Frequently Bought Together
Reagents for DEHA determination with Lovibond® MD photometers
: Lovibond
Congo red indicator
: Thermo Fisher Scientific
Sodium dodecyl sulphate (SDS) ≥99% (dried basis), HiPerSolv CHROMANORM® for ion pair chromatography
: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
: Lovibond
Reagents for Sulphide Determination using Lovibond® MD photometers
: Lovibond
NANOCOLOR® Photometric Tube Test Kit for Hydrocarbons
: MACHEREY-NAGEL