Stainless steel sealing machine with double sealing blade and adjustable sealing temperature. Max. width: 250mm Weight: 8kg 220Vac, 50Hz, 500W 1 * 1 items
Catalog # CERT8500483
Stainless steel sealing machine with double sealing blade and adjustable sealing temperature. Max. width: 250mm Weight: 8kg 220Vac, 50Hz, 500W 1 * 1 items
Catalog # CERT8500483
Frequently Bought Together