ContactUsLinkComponent
Slide Box Increase bundle for 30 mm microscope slides to enhance the height of GLW Slide Boxes to store slides up to 30 mm height. Suitable for GLW K50-Series and K51 Slide Boxes. Made of polycarbonate. 1 * 1 items
Catalog # GLWAZKH301
Supplier:  GLW
Slide Box Increase bundle for 30 mm microscope slides to enhance the height of GLW Slide Boxes to store slides up to 30 mm height. Suitable for GLW K50-Series and K51 Slide Boxes. Made of polycarbonate. 1 * 1 items
Catalog # GLWAZKH301
Supplier:  GLW
Frequently Bought Together