ContactUsLinkComponent
Tri-Clamp GL-Adaptor PTFE, Fitting for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Dimensions: Plate-Ø mm 25,5. Nominal size as per DIN DN 08, for tube I.D. mm 8,0. Connecting thread GL 14 1 * 1 items
Catalog # BOHLD901-02
Supplier:  Bohlender
undefined
Tri-Clamp GL-Adaptor PTFE, Fitting for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Dimensions: Plate-Ø mm 25,5. Nominal size as per DIN DN 08, for tube I.D. mm 8,0. Connecting thread GL 14 1 * 1 items
Catalog # BOHLD901-02
Supplier:  Bohlender
Frequently Bought Together