Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

ACETONE 1 * 20 L

Catalog # 2432-19
ACETONE 1 * 20 L
Catalog # 2432-19
Supplier:  
CAS Number:  
ACETONE 1 * 20 L
ACETONE 1 * 20 L
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Acetone Ph.Eur. 1 * 5 L Catalog #20165.367

 

Product Details & Documents