ContactUsLinkComponent
BCN, an alternative to DIBO and DBCO,  reacts with azide to form 1,2,3-triazole by copper free 1,3-dipolar Huisgen cycloaddition. MT S-BCN is a fast-reacting and highly selective thiol-reactive label that permits one to attach BCN to a thiol group. 1 * 1 mg
Catalog # BTIU96023
Supplier:  Biotium
BCN, an alternative to DIBO and DBCO,  reacts with azide to form 1,2,3-triazole by copper free 1,3-dipolar Huisgen cycloaddition. MT S-BCN is a fast-reacting and highly selective thiol-reactive label that permits one to attach BCN to a thiol group. 1 * 1 mg
Catalog # BTIU96023
Supplier:  Biotium
Frequently Bought Together