ContactUsLinkComponent
Drop-n-Stain™ CF543 goat anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.   To minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 5 mL
Catalog # BTIU20969
Supplier:  Biotium
Drop-n-Stain™ CF543 goat anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting.   To minimize cross-reactivity, the antibody has been adsorbed against human, bovine, horse, rabbit, and swine serum. 1 * 5 mL
Catalog # BTIU20969
Supplier:  Biotium
Frequently Bought Together