refractometer, rfm712-m, digital, 0-50 brix, 1 d.p, atc, keypad style, factory model 1 * 1 items
BELL19-00-61-182
refractometer, rfm712-m, digital, 0-50 brix, 1 d.p, atc, keypad style, factory model 1 * 1 items
BELL19-00-61-182
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection