ContactUsLinkComponent
Vacuum unit for MTP (Standard), modification to 10x reagent beakers (aluminium), 2000 ml, integrated vacuum function for two paraff in beakers, integrated vacuum function for ten reagent beakers (aluminium) 1 * 1 items
Catalog # SLEE11000225
Supplier:  Slee
Vacuum unit for MTP (Standard), modification to 10x reagent beakers (aluminium), 2000 ml, integrated vacuum function for two paraff in beakers, integrated vacuum function for ten reagent beakers (aluminium) 1 * 1 items
Catalog # SLEE11000225
Supplier:  Slee
Frequently Bought Together