ContactUsLinkComponent
Magnifier with illumination ø 72 mm lens, diopters : 6, rotatable and adjustable, light source : 63x Light-emitting diode (LED), neu tral white, dimmer, electrical parameters : 100-240 V 50/60 Hz 1 * 1 items
Catalog # SLEE10090060
Supplier:  Slee
Magnifier with illumination ø 72 mm lens, diopters : 6, rotatable and adjustable, light source : 63x Light-emitting diode (LED), neu tral white, dimmer, electrical parameters : 100-240 V 50/60 Hz 1 * 1 items
Catalog # SLEE10090060
Supplier:  Slee
Frequently Bought Together