ContactUsLinkComponent
Technical features: Electrical Single-phase Operation Vacuum Pressure: 2179 mm H2O Air Flow: 189 m /h Amperage: 5 A Hertz: 50 Hz Voltage: 240 V Power: 1050 W Bag Capacity: 19 L Dimensions: H64 cm x W46 cm x L76 cm Weight 30 Kg Noise Level: 75.00 dB ( 1 * 1 items
Catalog # TIGV110545B
Supplier:  TIGER-VAC
Technical features: Electrical Single-phase Operation Vacuum Pressure: 2179 mm H2O Air Flow: 189 m /h Amperage: 5 A Hertz: 50 Hz Voltage: 240 V Power: 1050 W Bag Capacity: 19 L Dimensions: H64 cm x W46 cm x L76 cm Weight 30 Kg Noise Level: 75.00 dB ( 1 * 1 items
Catalog # TIGV110545B
Supplier:  TIGER-VAC
Frequently Bought Together