ContactUsLinkComponent
Microplate 1536 well polystyrene white F-bottom HI-base non-binding non-sterile (15 pieces/bag) 1 * 60 items
Catalog # GREI782904_60
Supplier:  Greiner Bio-One
Microplate 1536 well polystyrene white F-bottom HI-base non-binding non-sterile (15 pieces/bag) 1 * 60 items
Catalog # GREI782904_60
Supplier:  Greiner Bio-One
Frequently Bought Together