PBS BUFFER (1X, DULBECCO'S) - POWDER 1 * 1 L
A0964.1000
Frequently Bought Together
VWR® Disposable Spatulas, PP
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Pasteur pipettes, disposable
: Brand
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection