ContactUsLinkComponent
Rabbit IgG polyclonal antibody for N-alpha-acetyltransferase 15, NatA auxiliary subunit(NAA15) detection. Tested with WB, IHC-P in H uman Mouse. 1 * 100 µG
Catalog # BSBTA05636-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for N-alpha-acetyltransferase 15, NatA auxiliary subunit(NAA15) detection. Tested with WB, IHC-P in H uman Mouse. 1 * 100 µG
Catalog # BSBTA05636-1
Supplier:  Boster Bio